Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014314.3 | NP_055129.2 | 6 - 7 | 956 | 126 |
GenBank mRNA | AF038963.1 | - | 6 - 7 | 955 | 126 |
AK125989.1 | - | 7 - 8 | 1161 | 126 | |
AK301678.1 | - | 5 - 6 | 679 | 126 | |
AK315040.1 | - | 6 - 7 | 827 | 126 | |
BC132786.1 | - | 6 - 7 | 904 | 126 | |
BC136610.1 | - | 6 - 7 | 904 | 126 | |
BX647917.1 | - | 5 - 6 | 693 | 126 |