Assay Design



Gene Transcripts

Gene Symbol
FYB
Entrez Gene ID
2533
Gene Name
FYN binding protein
Gene Aliases
ADAP, PRO0823, SLAP-130, SLAP130
Chromosome Location
Chr.5: 39105252 - 39274523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006714465.2 XP_006714528.1 2 - 3 479 79

View Full Product Details