Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001398.2 | NP_001389.2 | 2 - 3 | 344 | 70 |
XM_017026448.1 | XP_016881937.1 | 2 - 3 | 294 | 70 | |
GenBank mRNA | AK291860.1 | - | 2 - 3 | 279 | 70 |
AK301211.1 | - | 2 - 3 | 276 | 70 | |
BC011792.2 | - | 2 - 3 | 261 | 70 | |
BC017408.1 | - | 2 - 3 | 285 | 70 | |
BG527600.1 | - | 2 - 3 | 345 | 70 | |
U16660.1 | - | 2 - 3 | 286 | 70 |