Assay Design



Gene Transcripts

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
Chromosome Location
Chr.7: 100620416 - 100642780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005250553.4 XP_005250610.1 1 - 2 94 105

View Full Product Details