Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003258.4 | NP_003249.3 | 5 - 6 | 608 | 59 |
XM_017024992.1 | XP_016880481.1 | 5 - 6 | 612 | 59 | |
GenBank mRNA | AB451268.1 | - | 5 - 6 | 398 | 59 |
AB451395.1 | - | 5 - 6 | 398 | 59 | |
AK314950.1 | - | 5 - 6 | 451 | 59 | |
BC006484.1 | - | 5 - 6 | 438 | 59 | |
BC007872.2 | - | 5 - 6 | 425 | 59 | |
BC007986.2 | - | 5 - 6 | 434 | 59 | |
BI754079.1 | - | 5 - 6 | 639 | 59 | |
BT006941.1 | - | 5 - 6 | 398 | 59 | |
K02581.1 | - | 5 - 6 | 455 | 59 |