Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001992.1 | NP_001001992.1 | 10 - 11 | 1227 | 134 |
NM_001032410.1 | NP_001027582.1 | 11 - 12 | 1293 | 134 | |
NM_006447.2 | NP_006438.1 | 10 - 11 | 1230 | 134 | |
XM_017028257.1 | XP_016883746.1 | 11 - 12 | 1377 | 134 | |
XM_017028258.1 | XP_016883747.1 | 11 - 12 | 1256 | 134 | |
XM_017028259.1 | XP_016883748.1 | 11 - 12 | 1228 | 134 | |
XM_017028260.1 | XP_016883749.1 | 9 - 10 | 973 | 134 | |
XM_017028261.1 | XP_016883750.1 | 11 - 12 | 1220 | 134 | |
GenBank mRNA | AF113219.1 | - | 6 - 7 | 615 | 134 |
AF126736.1 | - | 10 - 11 | 1156 | 134 | |
AK023247.1 | - | 5 - 6 | 579 | 134 | |
AK025104.1 | - | 10 - 11 | 1193 | 134 | |
AK025989.1 | - | 10 - 11 | 1196 | 134 | |
AK222681.1 | - | 10 - 11 | 1159 | 134 | |
AK294284.1 | - | 9 - 10 | 1091 | 134 | |
AK302247.1 | - | 4 - 5 | 339 | 134 | |
AY333928.1 | - | 10 - 11 | 1156 | 134 | |
BC030777.2 | - | 10 - 11 | 1183 | 134 |