Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032410.1 | NP_001027582.1 | 6 - 7 | 714 | 150 |
NM_006447.2 | NP_006438.1 | 5 - 6 | 651 | 150 | |
XM_017028257.1 | XP_016883746.1 | 6 - 7 | 798 | 150 | |
XM_017028260.1 | XP_016883749.1 | 4 - 5 | 405 | 150 | |
GenBank mRNA | AF126736.1 | - | 5 - 6 | 577 | 150 |
AK222681.1 | - | 5 - 6 | 580 | 150 | |
AK222884.1 | - | 5 - 6 | 573 | 150 | |
AY333928.1 | - | 5 - 6 | 577 | 150 |