Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016513.4 | NP_057597.2 | 1 - 2 | 205 | 166 |
XM_011514421.1 | XP_011512723.1 | 1 - 2 | 175 | 166 | |
XM_017010486.1 | XP_016865975.1 | 1 - 2 | 223 | 166 | |
GenBank mRNA | AK074892.1 | - | 1 - 2 | 175 | 166 |