Assay Design
Gene Transcripts
- Gene Symbol
- PYY
- Entrez Gene ID
-
5697
- Gene Name
- peptide YY
- Gene Aliases
-
PYY-I, PYY1
- Chromosome Location
-
Chr.17: 43952733 - 44024260 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_004160.5 |
NP_004151.3 |
1 - 2 |
80 |
58 |
GenBank mRNA |
BC041057.1 |
- |
1 - 2 |
80 |
58 |
View Full Product Details