Assay Design



Gene Transcripts

Gene Symbol
PYY
Entrez Gene ID
5697
Gene Name
peptide YY
Gene Aliases
PYY-I, PYY1
Chromosome Location
Chr.17: 43952733 - 44024260 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004160.5 NP_004151.3 3 - 4 415 80
XM_011525035.1 XP_011523337.1 3 - 4 1025 80
GenBank mRNA BC041057.1 - 3 - 4 415 80

View Full Product Details