Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004774.3 | NP_004765.2 | 7 - 8 | 714 | 132 |
XM_005257465.3 | XP_005257522.1 | 6 - 7 | 588 | 132 | |
XM_006721957.1 | XP_006722020.1 | 7 - 8 | 711 | 132 | |
XM_017024779.1 | XP_016880268.1 | 7 - 8 | 721 | 132 | |
GenBank mRNA | AF055994.1 | - | 7 - 8 | 694 | 132 |
AF283812.1 | - | 6 - 7 | 457 | 132 | |
AK299789.1 | - | 6 - 7 | 579 | 132 | |
BC006517.2 | - | 7 - 8 | 687 | 132 | |
BC060758.1 | - | 7 - 8 | 692 | 132 | |
BC131783.1 | - | 7 - 8 | 568 | 132 | |
Y13467.1 | - | 7 - 8 | 692 | 132 |