Assay Design



Gene Transcripts

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
Chromosome Location
Chr.19: 19320686 - 19358754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015329.3 NP_056144.3 13 - 14 1354 77
XM_005259838.3 XP_005259895.3 11 - 12 1405 77
XM_011527844.2 XP_011526146.1 13 - 14 1759 77
GenBank mRNA AB020699.1 - 13 - 14 1340 77
AK025896.1 - 11 - 12 1006 77
AK128029.1 - 2 - 3 1212 77
BC010222.1 - 2 - 3 140 77

View Full Product Details