Assay Design



Gene Transcripts

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
Chromosome Location
Chr.19: 19320686 - 19358754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015329.3 NP_056144.3 3 - 4 398 62
XM_005259837.3 XP_005259894.2 3 - 4 390 62
XM_006722711.3 XP_006722774.2 3 - 4 390 62
XM_011527844.2 XP_011526146.1 3 - 4 803 62
GenBank mRNA AB020699.1 - 3 - 4 384 62
AK025896.1 - 1 - 2 50 62
BY795974.2 - 3 - 4 366 62

View Full Product Details