Assay Design



Gene Transcripts

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
Chromosome Location
Chr.19: 19320686 - 19358754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015329.3 NP_056144.3 7 - 8 777 71
XM_005259837.3 XP_005259894.2 7 - 8 769 71
XM_005259838.3 XP_005259895.3 5 - 6 828 71
XM_006722711.3 XP_006722774.2 7 - 8 769 71
XM_011527844.2 XP_011526146.1 7 - 8 1182 71
XM_017026539.1 XP_016882028.1 5 - 6 827 71
GenBank mRNA AB020699.1 - 7 - 8 763 71
AK025896.1 - 5 - 6 429 71

View Full Product Details