Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130105.1 | NP_001123577.1 | 15 - 16 | 2013 | 151 |
NM_005713.2 | NP_005704.1 | 14 - 15 | 1914 | 151 | |
NM_031361.2 | NP_112729.1 | 13 - 14 | 1836 | 151 | |
XM_006714513.2 | XP_006714576.1 | 14 - 15 | 1914 | 151 | |
XM_011543090.2 | XP_011541392.1 | 14 - 15 | 1914 | 151 | |
XM_011543091.2 | XP_011541393.1 | 13 - 14 | 1836 | 151 | |
XM_017008919.1 | XP_016864408.1 | 13 - 14 | 1836 | 151 | |
GenBank mRNA | AF136450.1 | - | 14 - 15 | 1900 | 151 |
AF232930.1 | - | 13 - 14 | 1804 | 151 | |
AK000604.1 | - | 9 - 10 | 856 | 151 | |
AK091851.1 | - | 14 - 15 | 1726 | 151 | |
AK096854.1 | - | 15 - 16 | 2170 | 151 | |
AK292087.1 | - | 13 - 14 | 1836 | 151 | |
AY453385.1 | - | 13 - 14 | 1414 | 151 | |
AY453386.1 | - | 14 - 15 | 1492 | 151 | |
BC000102.2 | - | 13 - 14 | 1820 | 151 |