Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130105.1 | NP_001123577.1 | 5 - 6 | 981 | 67 |
NM_005713.2 | NP_005704.1 | 4 - 5 | 882 | 67 | |
NM_031361.2 | NP_112729.1 | 4 - 5 | 882 | 67 | |
XM_006714513.2 | XP_006714576.1 | 4 - 5 | 882 | 67 | |
XM_011543090.2 | XP_011541392.1 | 4 - 5 | 882 | 67 | |
XM_011543091.2 | XP_011541393.1 | 4 - 5 | 882 | 67 | |
XM_017008919.1 | XP_016864408.1 | 4 - 5 | 882 | 67 | |
GenBank mRNA | AF136450.1 | - | 4 - 5 | 868 | 67 |
AF232930.1 | - | 4 - 5 | 850 | 67 | |
AK091851.1 | - | 5 - 6 | 772 | 67 | |
AK096854.1 | - | 5 - 6 | 1138 | 67 | |
AK292087.1 | - | 4 - 5 | 882 | 67 | |
AY453385.1 | - | 4 - 5 | 460 | 67 | |
AY453386.1 | - | 4 - 5 | 460 | 67 | |
BC000102.2 | - | 4 - 5 | 866 | 67 |