Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021156.3 | NP_066979.2 | 4 - 5 | 685 | 125 |
GenBank mRNA | AK074838.1 | - | 4 - 5 | 617 | 125 |
AK075404.1 | - | 4 - 5 | 664 | 125 | |
AK226005.1 | - | 3 - 4 | 219 | 125 | |
AL118494.1 | - | 3 - 4 | 210 | 125 | |
AY359004.1 | - | 4 - 5 | 602 | 125 | |
BC033787.1 | - | 4 - 5 | 496 | 125 | |
BC044777.1 | - | 4 - 5 | 685 | 125 |