Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021156.3 | NP_066979.2 | 7 - 8 | 895 | 80 |
GenBank mRNA | AK075404.1 | - | 7 - 8 | 874 | 80 |
AK226005.1 | - | 6 - 7 | 429 | 80 | |
AL118494.1 | - | 6 - 7 | 420 | 80 | |
AY359004.1 | - | 7 - 8 | 812 | 80 | |
BC033787.1 | - | 7 - 8 | 706 | 80 | |
BC044777.1 | - | 7 - 8 | 895 | 80 |