Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243081.1 | NP_001230010.1 | 3 - 4 | 434 | 76 |
NM_001422.3 | NP_001413.1 | 4 - 5 | 536 | 76 | |
NM_198381.1 | NP_938195.1 | 4 - 5 | 668 | 76 | |
GenBank mRNA | AF049703.1 | - | 4 - 5 | 517 | 76 |
AF115402.1 | - | 4 - 5 | 669 | 76 | |
AF115403.1 | - | 4 - 5 | 504 | 76 | |
AK075197.1 | - | 4 - 5 | 492 | 76 | |
AK290808.1 | - | 4 - 5 | 492 | 76 | |
AK290817.1 | - | 4 - 5 | 580 | 76 | |
AK312537.1 | - | 4 - 5 | 530 | 76 | |
BC029743.1 | - | 4 - 5 | 519 | 76 | |
BG260616.1 | - | - | 519 | 76 | |
BP358543.1 | - | 4 - 5 | 536 | 76 | |
DA630849.1 | - | 3 - 4 | 463 | 76 |