Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243080.1 | NP_001230009.1 | 3 - 4 | 324 | 80 |
NM_001243081.1 | NP_001230010.1 | 4 - 5 | 507 | 80 | |
NM_001422.3 | NP_001413.1 | 5 - 6 | 609 | 80 | |
NM_198381.1 | NP_938195.1 | 5 - 6 | 741 | 80 | |
XM_017017308.1 | XP_016872797.1 | 4 - 5 | 523 | 80 | |
GenBank mRNA | AF049703.1 | - | 5 - 6 | 590 | 80 |
AF115402.1 | - | 5 - 6 | 742 | 80 | |
AF115403.1 | - | 5 - 6 | 577 | 80 | |
AK075197.1 | - | 5 - 6 | 565 | 80 | |
AK290808.1 | - | 5 - 6 | 565 | 80 | |
AK290817.1 | - | 5 - 6 | 653 | 80 | |
AK312537.1 | - | 5 - 6 | 603 | 80 | |
BC029743.1 | - | 5 - 6 | 592 | 80 | |
DQ123839.1 | - | 3 - 4 | 329 | 80 |