Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031711.2 | NP_001026881.1 | 7 - 8 | 738 | 89 |
XM_006714892.1 | XP_006714955.1 | 5 - 6 | 429 | 89 | |
XM_011534597.1 | XP_011532899.1 | 7 - 8 | 631 | 89 | |
GenBank mRNA | AB033007.1 | - | 7 - 8 | 683 | 89 |
AF267855.1 | - | 5 - 6 | 637 | 89 | |
AK000752.1 | - | 1 - 2 | 59 | 89 | |
AK026049.1 | - | 4 - 5 | 391 | 89 | |
AK303453.1 | - | 4 - 5 | 549 | 89 | |
BC012766.2 | - | 7 - 8 | 620 | 89 | |
BC014490.1 | - | 7 - 8 | 613 | 89 |