Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318859.1 | NP_001305788.1 | 2 - 3 | 344 | 91 |
NM_080916.2 | NP_550438.1 | 2 - 3 | 344 | 91 | |
NM_080918.2 | NP_550440.1 | 2 - 3 | 344 | 91 | |
XM_011532647.2 | XP_011530949.1 | 2 - 3 | 340 | 91 | |
GenBank mRNA | BC015757.1 | - | 2 - 3 | 283 | 91 |
BU580854.1 | - | 2 - 3 | 328 | 91 | |
EL584040.1 | - | 2 - 3 | 308 | 91 | |
KU178023.1 | - | 2 - 3 | 259 | 91 | |
U41668.1 | - | 2 - 3 | 91 |