Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318860.1 | NP_001305789.1 | 2 - 3 | 417 | 155 |
NM_001318861.1 | NP_001305790.1 | 3 - 4 | 503 | 155 | |
NM_080916.2 | NP_550438.1 | 3 - 4 | 530 | 155 | |
NR_134894.1 | 2 - 3 | 417 | 155 | ||
GenBank mRNA | BC015757.1 | - | 3 - 4 | 469 | 155 |
BG470729.1 | - | 2 - 3 | 333 | 155 | |
BG575800.1 | - | 3 - 4 | 408 | 155 | |
BG708680.1 | - | 2 - 3 | 406 | 155 | |
BG715600.1 | - | 2 - 3 | 358 | 155 | |
KU178023.1 | - | 3 - 4 | 445 | 155 | |
KU178024.1 | - | 2 - 3 | 332 | 155 | |
U41668.1 | - | 3 - 4 | 479 | 155 |