Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015131.2 | NP_055946.1 | 12 - 13 | 1577 | 86 |
GenBank mRNA | AK000836.1 | - | 11 - 12 | 1273 | 86 |
D26488.1 | - | 12 - 13 | 1550 | 86 | |
D87716.1 | - | 10 - 11 | 1056 | 86 |