Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015131.2 | NP_055946.1 | 15 - 16 | 1795 | 114 |
GenBank mRNA | AK000836.1 | - | 14 - 15 | 1491 | 114 |
BC024234.1 | - | 1 - 2 | 111 | 114 | |
D26488.1 | - | 15 - 16 | 1768 | 114 | |
D87716.1 | - | 13 - 14 | 1274 | 114 |