Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015131.2 | NP_055946.1 | 5 - 6 | 798 | 140 |
GenBank mRNA | AK000836.1 | - | 4 - 5 | 494 | 140 |
BC033206.1 | - | 4 - 5 | 503 | 140 | |
D26488.1 | - | 5 - 6 | 771 | 140 | |
D87716.1 | - | 3 - 4 | 277 | 140 |