Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000922.3 | NP_000913.2 | 2 - 3 | 1384 | 102 |
XM_006718249.3 | XP_006718312.1 | 2 - 3 | 1416 | 102 | |
XM_017017911.1 | XP_016873400.1 | 2 - 3 | 1416 | 102 | |
XM_017017912.1 | XP_016873401.1 | 2 - 3 | 1416 | 102 | |
GenBank mRNA | AY459346.1 | - | 2 - 3 | 1322 | 102 |
BC136565.1 | - | 2 - 3 | 1277 | 102 | |
BC136566.1 | - | 2 - 3 | 1277 | 102 | |
BC144248.1 | - | 2 - 3 | 1277 | 102 | |
BC150307.1 | - | 2 - 3 | 1384 | 102 | |
X95520.1 | - | 2 - 3 | 1031 | 102 |