Assay Design
Gene Transcripts
- Gene Symbol
- SLC25A37
- Entrez Gene ID
-
51312
- Gene Name
- solute carrier family 25 member 37
- Gene Aliases
-
HT015, MFRN, MFRN1, MSC, MSCP, PRO1278, PRO1584, PRO2217
- Chromosome Location
-
Chr.8: 23528795 - 23575463 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF116640.1 |
- |
1 - 1 |
1160 |
138 |
AK093931.1 |
- |
1 - 1 |
1744 |
138 |
View Full Product Details