Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320666.1 | NP_001307595.1 | 5 - 6 | 840 | 124 |
NM_001320667.1 | NP_001307596.1 | 5 - 6 | 746 | 124 | |
NM_001320668.1 | NP_001307597.1 | 5 - 6 | 616 | 124 | |
NM_001320669.1 | NP_001307598.1 | 5 - 6 | 554 | 124 | |
NM_001320670.1 | NP_001307599.1 | 5 - 6 | 551 | 124 | |
NM_001320671.1 | NP_001307600.1 | 5 - 6 | 457 | 124 | |
NM_014898.2 | NP_055713.1 | 5 - 6 | 787 | 124 | |
GenBank mRNA | AB023178.1 | - | 5 - 6 | 787 | 124 |
AK297301.1 | - | 5 - 6 | 425 | 124 | |
BC041087.1 | - | 5 - 6 | 816 | 124 | |
BC050542.1 | - | 5 - 6 | 745 | 124 | |
GU388430.1 | - | 5 - 6 | 616 | 124 |