Assay Design



Gene Transcripts

Gene Symbol
PLEKHG2
Entrez Gene ID
64857
Gene Name
pleckstrin homology and RhoGEF domain containing G2
Gene Aliases
ARHGEF42, CLG, LDAMD
Chromosome Location
Chr.19: 39412582 - 39428415 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022835.2 NP_073746.2 12 - 13 1619 70
XM_005259163.2 XP_005259220.1 12 - 13 1440 70
XM_006723334.2 XP_006723397.1 12 - 13 1970 70
XM_011527232.2 XP_011525534.1 12 - 13 1546 70
XM_017027150.1 XP_016882639.1 12 - 13 2150 70
XM_017027151.1 XP_016882640.1 12 - 13 2150 70
GenBank mRNA AK024429.1 - 12 - 13 1619 70
AK026111.1 - 12 - 13 1428 70
AK302476.1 - 12 - 13 1243 70
BC013426.1 - 2 - 3 110 70
BC075858.1 - 10 - 11 984 70

View Full Product Details