Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000632.3 | NP_000623.2 | 17 - 18 | 2254 | 75 |
NM_001145808.1 | NP_001139280.1 | 17 - 18 | 2257 | 75 | |
XM_011545850.2 | XP_011544152.1 | 15 - 16 | 2053 | 75 | |
XM_017023216.1 | XP_016878705.1 | 17 - 18 | 2249 | 75 | |
GenBank mRNA | AK057856.1 | - | 5 - 6 | 640 | 75 |
AK127637.1 | - | 8 - 9 | 895 | 75 | |
BC096346.3 | - | 17 - 18 | 2220 | 75 | |
BC096347.3 | - | 17 - 18 | 2220 | 75 | |
BC096348.3 | - | 17 - 18 | 2220 | 75 | |
BC099660.3 | - | 17 - 18 | 2220 | 75 | |
J03925.1 | - | 17 - 18 | 2231 | 75 | |
J04145.1 | - | 16 - 17 | 2135 | 75 | |
M18044.1 | - | 17 - 18 | 2231 | 75 |