Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006662.2 | NP_006653.2 | 13 - 14 | 2382 | 71 |
GenBank mRNA | AB621816.1 | - | 13 - 14 | 2449 | 71 |
AF143946.1 | - | 9 - 10 | 1616 | 71 | |
AK023808.1 | - | 6 - 7 | 1096 | 71 | |
AK128030.1 | - | 13 - 14 | 2382 | 71 |