Assay Design



Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 15 - 16 1283 63
NM_001161358.1 NP_001154830.1 14 - 15 1193 63
NM_001161359.1 NP_001154831.1 13 - 14 1123 63
NM_015122.2 NP_055937.1 15 - 16 1287 63
XM_005259831.3 XP_005259888.1 14 - 15 1372 63
XM_006722701.3 XP_006722764.1 14 - 15 1291 63
XM_006722702.3 XP_006722765.1 14 - 15 1368 63
XM_011527824.2 XP_011526126.1 15 - 16 1283 63
XM_011527825.2 XP_011526127.1 15 - 16 1272 63
XM_011527826.2 XP_011526128.1 15 - 16 1255 63
XM_011527827.2 XP_011526129.1 13 - 14 1090 63
XM_011527828.2 XP_011526130.1 14 - 15 1186 63
XM_017026519.1 XP_016882008.1 15 - 16 1353 63
XM_017026520.1 XP_016882009.1 15 - 16 1336 63
XM_017026521.1 XP_016882010.1 14 - 15 1287 63
XM_017026522.1 XP_016882011.1 15 - 16 1283 63
XM_017026523.1 XP_016882012.1 15 - 16 1353 63
XM_017026524.1 XP_016882013.1 14 - 15 1368 63
XM_017026525.1 XP_016882014.1 14 - 15 1166 63
XM_017026526.1 XP_016882015.1 15 - 16 1283 63
XM_017026527.1 XP_016882016.1 14 - 15 1291 63
XM_017026528.1 XP_016882017.1 15 - 16 1353 63
GenBank mRNA AB006628.1 - 12 - 13 1057 63
AK303623.1 - 11 - 12 985 63
AK309403.1 - 11 - 12 1036 63
AK316458.1 - 13 - 14 1094 63
BC028021.1 - 15 - 16 1228 63
BC041130.1 - 15 - 16 1237 63

View Full Product Details