Assay Design



Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 21 - 22 2021 75
NM_001161358.1 NP_001154830.1 20 - 21 1931 75
NM_001161359.1 NP_001154831.1 19 - 20 1861 75
NM_015122.2 NP_055937.1 21 - 22 2025 75
XM_005259831.3 XP_005259888.1 20 - 21 2110 75
XM_006722701.3 XP_006722764.1 20 - 21 2029 75
XM_006722702.3 XP_006722765.1 20 - 21 2106 75
XM_011527824.2 XP_011526126.1 21 - 22 2021 75
XM_011527825.2 XP_011526127.1 21 - 22 2010 75
XM_011527826.2 XP_011526128.1 21 - 22 1993 75
XM_011527827.2 XP_011526129.1 18 - 19 1552 75
XM_011527828.2 XP_011526130.1 20 - 21 1924 75
XM_017026519.1 XP_016882008.1 21 - 22 2091 75
XM_017026520.1 XP_016882009.1 21 - 22 2074 75
XM_017026521.1 XP_016882010.1 20 - 21 2025 75
XM_017026522.1 XP_016882011.1 21 - 22 1904 75
XM_017026523.1 XP_016882012.1 21 - 22 1974 75
XM_017026524.1 XP_016882013.1 20 - 21 1989 75
XM_017026525.1 XP_016882014.1 20 - 21 1787 75
XM_017026526.1 XP_016882015.1 20 - 21 1745 75
XM_017026527.1 XP_016882016.1 19 - 20 1753 75
XM_017026528.1 XP_016882017.1 20 - 21 1815 75
GenBank mRNA AB006628.1 - 18 - 19 1795 75
AK291410.1 - 20 - 21 1887 75
AK303623.1 - 17 - 18 1723 75
AK309403.1 - 17 - 18 1774 75
AK316458.1 - 19 - 20 1832 75
BC028021.1 - 21 - 22 1966 75
BC041130.1 - 21 - 22 1975 75

View Full Product Details