Assay Design



Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 27 - 28 2760 95
NM_001161358.1 NP_001154830.1 26 - 27 2670 95
NM_001161359.1 NP_001154831.1 25 - 26 2600 95
NM_015122.2 NP_055937.1 27 - 28 2764 95
XM_005259831.3 XP_005259888.1 26 - 27 2849 95
XM_006722701.3 XP_006722764.1 26 - 27 2768 95
XM_006722702.3 XP_006722765.1 26 - 27 2845 95
XM_011527824.2 XP_011526126.1 27 - 28 2760 95
XM_011527825.2 XP_011526127.1 27 - 28 2749 95
XM_011527826.2 XP_011526128.1 27 - 28 2732 95
XM_011527827.2 XP_011526129.1 24 - 25 2291 95
XM_011527828.2 XP_011526130.1 26 - 27 2663 95
XM_017026519.1 XP_016882008.1 27 - 28 2830 95
XM_017026520.1 XP_016882009.1 27 - 28 2813 95
XM_017026521.1 XP_016882010.1 26 - 27 2764 95
XM_017026522.1 XP_016882011.1 27 - 28 2643 95
XM_017026523.1 XP_016882012.1 27 - 28 2713 95
XM_017026524.1 XP_016882013.1 26 - 27 2728 95
XM_017026525.1 XP_016882014.1 26 - 27 2526 95
XM_017026526.1 XP_016882015.1 26 - 27 2484 95
XM_017026527.1 XP_016882016.1 25 - 26 2492 95
XM_017026528.1 XP_016882017.1 26 - 27 2554 95
GenBank mRNA AB006628.1 - 24 - 25 2534 95
AK291410.1 - 26 - 27 2626 95
AK303623.1 - 23 - 24 2462 95
AK309403.1 - 23 - 24 2513 95
AK316458.1 - 25 - 26 2571 95
BC028021.1 - 27 - 28 2705 95
BC041130.1 - 27 - 28 2714 95

View Full Product Details