Assay Design



Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 8 - 9 774 57
NM_001161358.1 NP_001154830.1 7 - 8 684 57
NM_001161359.1 NP_001154831.1 6 - 7 614 57
NM_015122.2 NP_055937.1 8 - 9 778 57
XM_005259831.3 XP_005259888.1 7 - 8 859 57
XM_006722701.3 XP_006722764.1 7 - 8 782 57
XM_006722702.3 XP_006722765.1 7 - 8 859 57
XM_011527824.2 XP_011526126.1 8 - 9 774 57
XM_011527825.2 XP_011526127.1 8 - 9 763 57
XM_011527826.2 XP_011526128.1 8 - 9 746 57
XM_011527827.2 XP_011526129.1 6 - 7 581 57
XM_011527828.2 XP_011526130.1 7 - 8 677 57
XM_017026519.1 XP_016882008.1 8 - 9 844 57
XM_017026520.1 XP_016882009.1 8 - 9 827 57
XM_017026521.1 XP_016882010.1 7 - 8 778 57
XM_017026522.1 XP_016882011.1 8 - 9 774 57
XM_017026523.1 XP_016882012.1 8 - 9 844 57
XM_017026524.1 XP_016882013.1 7 - 8 859 57
XM_017026525.1 XP_016882014.1 7 - 8 657 57
XM_017026526.1 XP_016882015.1 8 - 9 774 57
XM_017026527.1 XP_016882016.1 7 - 8 782 57
XM_017026528.1 XP_016882017.1 8 - 9 844 57
GenBank mRNA AB006628.1 - 5 - 6 548 57
AK291410.1 - 7 - 8 640 57
AK303623.1 - 4 - 5 476 57
AK309403.1 - 4 - 5 527 57
AK316458.1 - 6 - 7 585 57
BC028021.1 - 8 - 9 719 57
BC041130.1 - 8 - 9 728 57

View Full Product Details