Assay Design



Gene Transcripts

Gene Symbol
MUC16
Entrez Gene ID
94025
Gene Name
mucin 16, cell surface associated
Gene Aliases
CA125
Chromosome Location
Chr.19: 8848840 - 9010390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024690.2 NP_078966.2 61 - 62 41459 62
XM_017027486.1 XP_016882975.1 55 - 56 40539 62
XM_017027487.1 XP_016882976.1 54 - 55 40473 62
XM_017027488.1 XP_016882977.1 55 - 56 40539 62
XM_017027489.1 XP_016882978.1 52 - 53 40385 62
XM_017027490.1 XP_016882979.1 50 - 51 40074 62
XM_017027492.1 XP_016882981.1 50 - 51 40071 62
XM_017027493.1 XP_016882982.1 50 - 51 40071 62
XM_017027494.1 XP_016882983.1 50 - 51 40071 62
XM_017027495.1 XP_016882984.1 50 - 51 40071 62
XM_017027497.1 XP_016882986.1 30 - 31 38196 62
XM_017027499.1 XP_016882988.1 55 - 56 31326 62
XM_017027500.1 XP_016882989.1 54 - 55 31245 62
XM_017027501.1 XP_016882990.1 34 - 35 3253 62
GenBank mRNA AF361486.3 - - 18719 62
AF414442.2 - - 64394 62
AK024365.1 - 13 - 14 1202 62
AK128681.1 - 11 - 12 1838 62
AK128856.1 - 18 - 19 1879 62
AK128857.1 - - 3155 62

View Full Product Details