Assay Design



Gene Transcripts

Gene Symbol
DHX40
Entrez Gene ID
79665
Gene Name
DEAH-box helicase 40
Gene Aliases
ARG147, DDX40, PAD
Chromosome Location
Chr.17: 59565525 - 59608352 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017025082.1 XP_016880571.1 2 - 3 389 147
XM_017025083.1 XP_016880572.1 2 - 3 386 147

View Full Product Details