Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314018.1 | NP_001300947.1 | 3 - 4 | 243 | 68 |
NM_020939.1 | NP_065990.1 | 14 - 15 | 1040 | 68 | |
XM_005249247.2 | XP_005249304.1 | 15 - 16 | 1091 | 68 | |
XM_011514768.1 | XP_011513070.1 | 15 - 16 | 1091 | 68 | |
XM_011514769.1 | XP_011513071.1 | 14 - 15 | 1040 | 68 | |
XM_011514770.1 | XP_011513072.1 | 16 - 17 | 1106 | 68 | |
XM_011514771.2 | XP_011513073.1 | 15 - 16 | 1431 | 68 | |
XM_011514772.1 | XP_011513074.1 | 15 - 16 | 1091 | 68 | |
XM_011514773.2 | XP_011513075.1 | 7 - 8 | 520 | 68 | |
XM_011514775.2 | XP_011513077.1 | 3 - 4 | 234 | 68 | |
XM_017011139.1 | XP_016866628.1 | 13 - 14 | 961 | 68 | |
GenBank mRNA | AB046819.1 | - | 14 - 15 | 1598 | 68 |
BC053872.1 | - | 3 - 4 | 243 | 68 |