Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314018.1 | NP_001300947.1 | 4 - 5 | 288 | 60 |
NM_001314019.1 | NP_001300948.1 | 5 - 6 | 329 | 60 | |
NM_001314020.1 | NP_001300949.1 | 3 - 4 | 226 | 60 | |
NM_020939.1 | NP_065990.1 | 15 - 16 | 1085 | 60 | |
XM_005249247.2 | XP_005249304.1 | 16 - 17 | 1136 | 60 | |
XM_011514768.1 | XP_011513070.1 | 16 - 17 | 1136 | 60 | |
XM_011514769.1 | XP_011513071.1 | 15 - 16 | 1085 | 60 | |
XM_011514770.1 | XP_011513072.1 | 17 - 18 | 1151 | 60 | |
XM_011514771.2 | XP_011513073.1 | 16 - 17 | 1476 | 60 | |
XM_011514772.1 | XP_011513074.1 | 16 - 17 | 1136 | 60 | |
XM_011514773.2 | XP_011513075.1 | 8 - 9 | 565 | 60 | |
XM_011514775.2 | XP_011513077.1 | 4 - 5 | 279 | 60 | |
XM_017011139.1 | XP_016866628.1 | 14 - 15 | 1006 | 60 | |
GenBank mRNA | AB046819.1 | - | 15 - 16 | 1643 | 60 |
BC053872.1 | - | 4 - 5 | 288 | 60 |