Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020939.1 | NP_065990.1 | 12 - 13 | 926 | 84 |
XM_005249247.2 | XP_005249304.1 | 13 - 14 | 977 | 84 | |
XM_011514768.1 | XP_011513070.1 | 13 - 14 | 977 | 84 | |
XM_011514769.1 | XP_011513071.1 | 12 - 13 | 926 | 84 | |
XM_011514770.1 | XP_011513072.1 | 14 - 15 | 992 | 84 | |
XM_011514771.2 | XP_011513073.1 | 13 - 14 | 1317 | 84 | |
XM_011514772.1 | XP_011513074.1 | 13 - 14 | 977 | 84 | |
XM_011514773.2 | XP_011513075.1 | 5 - 6 | 406 | 84 | |
XM_017011139.1 | XP_016866628.1 | 11 - 12 | 847 | 84 | |
GenBank mRNA | AB046819.1 | - | 12 - 13 | 1484 | 84 |