Assay Design



Gene Transcripts

Gene Symbol
UPP1
Entrez Gene ID
7378
Gene Name
uridine phosphorylase 1
Gene Aliases
UDRPASE, UP, UPASE, UPP
Chromosome Location
Chr.7: 48088308 - 48108746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287426.1 NP_001274355.1 1 - 2 323 90
GenBank mRNA BC007348.2 - 1 - 2 282 90

View Full Product Details