Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318323.1 | NP_001305252.1 | 5 - 5 | 2717 | 82 |
NM_145912.6 | NP_666017.1 | 6 - 6 | 2830 | 82 | |
XM_005261374.4 | XP_005261431.1 | 8 - 8 | 3457 | 82 | |
GenBank mRNA | AK074094.1 | - | 5 - 5 | 2422 | 82 |
AK097971.1 | - | 1 - 1 | 1783 | 82 | |
BC038241.1 | - | 6 - 6 | 2825 | 82 |