Assay Design



Gene Transcripts

Gene Symbol
MREG
Entrez Gene ID
55686
Gene Name
melanoregulin
Gene Aliases
DSU, WDT2
Chromosome Location
Chr.2: 215939097 - 216013623 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018000.2 NP_060470.2 3 - 4 641 94
XM_011511468.2 XP_011509770.1 4 - 5 641 94
GenBank mRNA AF068290.1 - 2 - 3 140 94
AK000978.1 - 3 - 4 611 94
BC032747.1 - 3 - 4 650 94
BC082990.1 - 3 - 4 631 94
CX781801.1 - 3 - 4 636 94

View Full Product Details