Assay Design



Gene Transcripts

Gene Symbol
MFF
Entrez Gene ID
56947
Gene Name
mitochondrial fission factor
Gene Aliases
C2orf33, GL004
Chromosome Location
Chr.2: 227325151 - 227357836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277061.1 NP_001263990.1 10 - 11 1419 92
NM_001277062.1 NP_001263991.1 8 - 9 1166 92
NM_001277063.1 NP_001263992.1 7 - 8 1022 92
NM_001277064.1 NP_001263993.1 7 - 8 1007 92
NM_001277065.1 NP_001263994.1 6 - 7 947 92
NM_001277066.1 NP_001263995.1 5 - 6 835 92
NM_001277067.1 NP_001263996.1 7 - 8 852 92
NM_001277068.1 NP_001263997.1 7 - 8 772 92
NM_020194.5 NP_064579.3 9 - 10 1156 92
NR_102266.1 4 - 5 614 92
XM_005246681.3 XP_005246738.1 9 - 10 1279 92
XM_005246682.1 XP_005246739.1 9 - 10 1264 92
XM_005246683.1 XP_005246740.1 9 - 10 1161 92
XM_005246684.1 XP_005246741.1 8 - 9 1204 92
XM_005246685.3 XP_005246742.1 9 - 10 1180 92
XM_005246686.3 XP_005246743.1 8 - 9 1120 92
XM_005246687.3 XP_005246744.1 8 - 9 1105 92
XM_005246688.1 XP_005246745.1 7 - 8 1045 92
XM_006712637.1 XP_006712700.1 8 - 9 1063 92
XM_006712638.1 XP_006712701.1 7 - 8 988 92
XM_006712639.2 XP_006712702.1 6 - 7 829 92
XM_011511500.1 XP_011509802.1 10 - 11 1211 92
XM_011511501.1 XP_011509803.1 9 - 10 1232 92
XM_017004512.1 XP_016860001.1 9 - 10 1328 92
XM_017004513.1 XP_016860002.1 7 - 8 1026 92
XM_017004514.1 XP_016860003.1 6 - 7 928 92
XM_017004515.1 XP_016860004.1 6 - 7 844 92
GenBank mRNA AF226049.1 - 5 - 6 590 92
AF246239.1 - 9 - 10 1156 92
AF258660.1 - 10 - 11 1274 92
AK026137.1 - 8 - 9 1067 92
AL833032.1 - 3 - 4 796 92
BC000797.1 - 6 - 7 679 92
BC093024.1 - 7 - 8 905 92
BQ653061.1 - 5 - 6 577 92

View Full Product Details