Assay Design



Gene Transcripts

Gene Symbol
GPR35
Entrez Gene ID
2859
Gene Name
G protein-coupled receptor 35
Gene Aliases
-
Chromosome Location
Chr.2: 240605408 - 240631259 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF089087.1 - 1 - 1 386 103
AK131540.1 - 1 - 1 2322 103
BC117453.1 - 1 - 1 183 103
BC117455.1 - 1 - 1 181 103

View Full Product Details