Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003852.3 | NP_003843.3 | 5 - 6 | 1089 | 145 |
NM_015905.2 | NP_056989.2 | 5 - 6 | 1089 | 145 | |
GenBank mRNA | AF009353.1 | - | 5 - 6 | 862 | 145 |
AF119042.1 | - | 5 - 6 | 990 | 145 | |
AK075306.1 | - | 5 - 6 | 1089 | 145 | |
AK302679.1 | - | 6 - 7 | 962 | 145 | |
AK312610.1 | - | 5 - 6 | 1217 | 145 | |
BC028689.2 | - | 5 - 6 | 987 | 145 |