Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015905.2 | NP_056989.2 | 9 - 10 | 1745 | 91 |
GenBank mRNA | AK075306.1 | - | 9 - 10 | 1745 | 91 |
AK302679.1 | - | 10 - 11 | 1618 | 91 | |
BC028689.2 | - | 9 - 10 | 1643 | 91 |