Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145337.2 | NP_001138809.1 | 3 - 4 | 218 | 150 |
NM_001145339.2 | NP_001138811.1 | 3 - 4 | 468 | 150 | |
NM_002392.5 | NP_002383.2 | 3 - 4 | 468 | 150 | |
XM_005268872.4 | XP_005268929.1 | 3 - 4 | 396 | 150 | |
GenBank mRNA | AF092843.1 | - | 3 - 4 | 176 | 150 |
AF092844.1 | - | 3 - 4 | 176 | 150 | |
AF385323.1 | - | 2 - 3 | 144 | 150 | |
AJ430612.1 | - | 2 - 3 | 144 | 150 | |
AJ430614.1 | - | 2 - 3 | 144 | 150 | |
AJ491698.1 | - | 2 - 3 | 144 | 150 | |
AJ491700.1 | - | 2 - 3 | 144 | 150 | |
AK290341.1 | - | 3 - 4 | 418 | 150 | |
AM902646.1 | - | 2 - 3 | 144 | 150 | |
AM902650.1 | - | 2 - 3 | 144 | 150 | |
AM902651.1 | - | 2 - 3 | 144 | 150 | |
AM902654.1 | - | 2 - 3 | 144 | 150 | |
AM902655.1 | - | 2 - 3 | 144 | 150 | |
AM902659.1 | - | 2 - 3 | 144 | 150 | |
AM902660.1 | - | 2 - 3 | 144 | 150 | |
AM902666.1 | - | 2 - 3 | 144 | 150 | |
AM902667.1 | - | 2 - 3 | 144 | 150 | |
AM902668.1 | - | 2 - 3 | 144 | 150 | |
AM902669.1 | - | 2 - 3 | 144 | 150 | |
AM902672.1 | - | 2 - 3 | 144 | 150 | |
AM902678.1 | - | 2 - 3 | 144 | 150 | |
AM902680.1 | - | 2 - 3 | 144 | 150 | |
AM902681.1 | - | 2 - 3 | 144 | 150 | |
BC152384.1 | - | 2 - 3 | 147 | 150 | |
BC152390.1 | - | 2 - 3 | 147 | 150 | |
BE296905.1 | - | 3 - 4 | 433 | 150 | |
BT007258.1 | - | 2 - 3 | 144 | 150 | |
EU076749.1 | - | 3 - 4 | 218 | 150 | |
HM213901.1 | - | 2 - 3 | 144 | 150 | |
JX015379.1 | - | 2 - 3 | 144 | 150 | |
JX015390.1 | - | 2 - 3 | 144 | 150 | |
JX015392.1 | - | 2 - 3 | 144 | 150 | |
M92424.1 | - | 3 - 4 | 455 | 150 | |
U33203.1 | - | 2 - 3 | 144 | 150 | |
Z12020.1 | - | 3 - 4 | 455 | 150 |