Assay Design



Gene Transcripts

Gene Symbol
MDM2
Entrez Gene ID
4193
Gene Name
MDM2 proto-oncogene
Gene Aliases
ACTFS, HDMX, hdm2
Chromosome Location
Chr.12: 68808149 - 68845544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145339.2 NP_001138811.1 7 - 8 972 149
NM_001278462.1 NP_001265391.1 4 - 5 377 149
NM_002392.5 NP_002383.2 9 - 10 1137 149
XM_005268872.4 XP_005268929.1 9 - 10 1065 149
XM_006719399.3 XP_006719462.1 7 - 8 720 149
XM_006719400.3 XP_006719463.1 8 - 9 1026 149
GenBank mRNA AF092844.1 - 7 - 8 680 149
AF092845.1 - 8 - 9 710 149
AF385325.1 - 4 - 5 394 149
AF385327.1 - 3 - 4 303 149
AK290341.1 - 9 - 10 1087 149
AM902642.1 - 9 - 10 906 149
AM902643.1 - 8 - 9 705 149
AM902644.1 - 7 - 8 679 149
AM902645.1 - 3 - 4 389 149
AM902647.1 - 8 - 9 739 149
AM902648.1 - 7 - 8 676 149
AM902649.1 - 4 - 5 464 149
AM902653.1 - 5 - 6 443 149
AM902655.1 - 5 - 6 487 149
AM902656.1 - 8 - 9 722 149
AM902657.1 - 4 - 5 353 149
AM902658.1 - 9 - 10 900 149
AM902659.1 - 8 - 9 731 149
AM902661.1 - 7 - 8 679 149
AM902662.1 - 6 - 7 604 149
AM902663.1 - 5 - 6 514 149
AM902664.1 - 6 - 7 629 149
AM902665.1 - 8 - 9 724 149
AM902666.1 - 8 - 9 697 149
AM902667.1 - 8 - 9 813 149
AM902669.1 - 5 - 6 598 149
AM902670.1 - 7 - 8 738 149
AM902671.1 - 5 - 6 554 149
AM902681.1 - 8 - 9 752 149
BC152384.1 - 8 - 9 816 149
BC152390.1 - 8 - 9 816 149
BT007258.1 - 8 - 9 813 149
EU076748.1 - 4 - 5 377 149
GQ848196.1 - 7 - 8 738 149
HM213901.1 - 7 - 8 652 149
HM213902.1 - 6 - 7 518 149
HM213903.1 - 4 - 5 464 149
M92424.1 - 9 - 10 1124 149
U33199.1 - 2 - 3 228 149
U33201.1 - 3 - 4 303 149
Z12020.1 - 9 - 10 1124 149

View Full Product Details